Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
FCER1G
Entrez Gene ID
397406
Gene Name
Fc fragment of IgE receptor Ig
Gene Aliases
-
Chromosome Location
Chr.4: 89242467 - 89246344 on Build Sscrofa11.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001001265.1 NP_001001265.1 2 - 3 190 77
GenBank mRNA AF148221.1 - 2 - 3 190 77
AK230637.1 - 2 - 3 219 77
AK230931.1 - 2 - 3 239 77
AK231016.1 - 2 - 3 239 77
AK231043.1 - 2 - 3 213 77
AK231062.1 - 2 - 3 266 77
AK231092.1 - 2 - 3 232 77
AK231129.1 - 2 - 3 213 77
AK343716.1 - 2 - 3 77
AK343723.1 - 2 - 3 213 77
AK343862.1 - 2 - 3 213 77
AK343884.1 - 2 - 3 213 77
AK343896.1 - 2 - 3 213 77
AK343918.1 - 2 - 3 232 77
AK343960.1 - 2 - 3 213 77
AK343964.1 - 2 - 3 232 77

View Full Product Details