Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
CFLAR
Entrez Gene ID
414381
Gene Name
CASP8 and FADD like apoptosis regulator
Gene Aliases
C-FLIP, FLIP
Chromosome Location
Chr.15: 104780744 - 104835631 on Build Sscrofa11.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001001628.1 NP_001001628.1 2 - 3 507 97
XM_005652441.3 XP_005652498.1 - 1592 97
XM_005652443.3 XP_005652500.1 - 1421 97
XM_005652444.3 XP_005652501.1 - 1811 97
XM_005652445.3 XP_005652502.1 - 1180 97
XM_005652446.3 XP_005652503.1 - 849 97
XM_005652447.3 XP_005652504.1 - 1009 97
XM_013996333.2 XP_013851787.1 - 2336 97
XM_013996365.2 XP_013851819.1 - 646 97
XM_021074699.1 XP_020930358.1 - 2541 97
GenBank mRNA AB370266.1 - 4 - 5 808 97
AB370267.1 - 4 - 5 808 97
AK234664.1 - 4 - 5 807 97
AK235528.1 - 3 - 4 823 97
AK238641.1 - 2 - 3 307 97
AK389814.1 - 3 - 4 577 97
AK395023.1 - 3 - 4 707 97
AK396797.1 - 3 - 4 608 97
AY533020.1 - 2 - 3 507 97
AY533021.1 - 2 - 3 507 97

View Full Product Details