Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
NFKBIA
Entrez Gene ID
406188
Gene Name
NFKB inhibitor alpha
Gene Aliases
ECI-6/IKBA, IKBA
Chromosome Location
Chr.7: 64508119 - 64510754 on Build Sscrofa11.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001005150.1 NP_001005150.1 4 - 5 632 54
GenBank mRNA AK233732.1 - 3 - 4 396 54
AK239662.1 - 5 - 6 892 54
AK389460.1 - 3 - 4 1027 54
AK395569.1 - 4 - 5 1008 54
AK395997.1 - 3 - 4 1094 54
AK397107.1 - 4 - 5 778 54
AK398938.1 - 4 - 5 743 54
AK399095.1 - 3 - 4 637 54
AK399188.1 - 4 - 5 743 54
AY609599.1 - 4 - 5 729 54
FN421467.1 - 4 - 5 569 54

View Full Product Details