The assay is designed to a sequence that does not exhibit typical exon-intron-exon gene structure, such as a fusion transcript, or an artificial, prokaryotic or mitochondrial sequence.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001011508.2 | NP_001011508.1 | 7 - 8 | 1671 | 77 |
XM_021077323.1 | XP_020932982.1 | - | 1640 | 77 | |
XM_021077324.1 | XP_020932983.1 | - | 1309 | 77 | |
GenBank mRNA | AJ544724.1 | - | 7 - 8 | 1671 | 77 |
KJ820675.1 | - | 7 - 8 | 1671 | 77 |