Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
KLF4
Entrez Gene ID
595111
Gene Name
Kruppel like factor 4
Gene Aliases
EZF, GKLF
Chromosome Location
Chr.1: 248604817 - 248609574 on Build Sscrofa11.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001031782.2 NP_001026952.2 4 - 5 1884 85
XM_005660316.3 XP_005660373.1 - 1586 85
GenBank mRNA AK397440.1 - 2 - 3 972 85
DQ000310.1 - 2 - 3 1361 85
EU669074.2 - 4 - 5 1884 85
EU669075.2 - 3 - 4 1983 85

View Full Product Details