Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
PLIN1
Entrez Gene ID
654411
Gene Name
perilipin 1
Gene Aliases
PLIN
Chromosome Location
Chr.7: 55237517 - 55250797 on Build Sscrofa11.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001038638.1 NP_001033727.1 6 - 7 987 164
GenBank mRNA AY973170.1 - 6 - 7 987 164

View Full Product Details