Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
BPIFA2
Entrez Gene ID
396649
Gene Name
BPI fold containing family A member 2
Gene Aliases
PSP
Chromosome Location
Chr.17: 36690008 - 36699989 on Build Sscrofa11.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_213767.2 NP_998932.1 5 - 6 648 97
GenBank mRNA AK350991.1 - 5 - 6 95
AY205234.1 - 5 - 6 638 97

View Full Product Details