Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
DSPP
Entrez Gene ID
396664
Gene Name
dentin sialophosphoprotein
Gene Aliases
DPP, DSP
Chromosome Location
Chr.8: 131318187 - 131444274 on Build Sscrofa11.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_021100402.1 XP_020956061.1 - 707 93
XM_021100403.1 XP_020956062.1 - 622 93
XM_021100404.1 XP_020956063.1 - 480 93
XM_021100405.1 XP_020956064.1 - 389 93
XM_021100406.1 XP_020956065.1 - 702 93
GenBank mRNA AF332578.1 - 3 - 4 233 93
AY161862.1 - 3 - 4 231 93
AY161863.1 - 3 - 4 241 93

View Full Product Details