Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
SERPINA6
Entrez Gene ID
396736
Gene Name
serpin family A member 6
Gene Aliases
CBG
Chromosome Location
Chr.7: 115535552 - 115554536 on Build Sscrofa11.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_213812.1 NP_998977.1 3 - 4 976 60
XM_013978394.2 XP_013833848.1 - 1332 60
XM_013978395.2 XP_013833849.1 - 1228 60
GenBank mRNA AF324155.1 - 3 - 4 976 60
AF506699.1 - 1 - 2 206 60
AK232280.1 - 3 - 4 981 60
AK233104.1 - 3 - 4 979 60
AK394279.1 - 3 - 4 1034 60

View Full Product Details