Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
GPX5
Entrez Gene ID
396920
Gene Name
glutathione peroxidase 5
Gene Aliases
-
Chromosome Location
Chr.7: 22289432 - 22300727 on Build Sscrofa11.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_213886.1 NP_999051.1 1 - 2 104 122
GenBank mRNA D37916.1 - 1 - 2 104 122

View Full Product Details