Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
DAO
Entrez Gene ID
397134
Gene Name
D-amino acid oxidase
Gene Aliases
DAAO, DAMOX, DAO1
Chromosome Location
Chr.14: 41931549 - 41950628 on Build Sscrofa11.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_214066.2 NP_999231.1 9 - 10 1025 69
GenBank mRNA AK390956.1 - 9 - 10 1007 69
AK391151.1 - 9 - 10 898 69
M16972.1 - 9 - 10 1025 69
M18448.1 - 4 - 5 358 69

View Full Product Details