Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
PLIN2
Entrez Gene ID
397402
Gene Name
perilipin 2
Gene Aliases
ADFP, ADRP
Chromosome Location
Chr.1: 203683867 - 203709277 on Build Sscrofa11.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_214200.2 NP_999365.1 5 - 6 668 112
XM_005660074.3 XP_005660131.1 - 705 112
XM_005660075.3 XP_005660132.1 - 848 112
XM_013993524.2 XP_013848978.1 - 759 112
XM_021082975.1 XP_020938634.1 - 704 112
GenBank mRNA AB527057.1 - 4 - 5 593 112
AF234676.2 - 5 - 6 669 112
AK232656.1 - 5 - 6 668 112
AY550037.1 - 5 - 6 666 112
DQ066659.1 - 4 - 5 593 112
JN616387.1 - 4 - 5 593 112
KC733245.1 - 4 - 5 593 112

View Full Product Details