Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
FABP6
Entrez Gene ID
397423
Gene Name
fatty acid binding protein 6
Gene Aliases
GT, ILBP, ILLBP, PIP
Chromosome Location
Chr.16: 63297594 - 63303675 on Build Sscrofa11.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_214215.2 NP_999380.2 3 - 4 399 83
GenBank mRNA AK231219.1 - 3 - 4 387 83
BW983662.1 - 3 - 4 386 83
EW435209.2 - 3 - 4 425 83
J05136.1 - 4 - 5 429 83

View Full Product Details