Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
IMPA1
Entrez Gene ID
397425
Gene Name
inositol monophosphatase 1
Gene Aliases
-
Chromosome Location
Chr.4: 54935838 - 54968672 on Build Sscrofa11.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_214216.1 NP_999381.1 6 - 7 567 117
XM_021088713.1 XP_020944372.1 - 682 117
XM_021088714.1 XP_020944373.1 - 734 117
XM_021088715.1 XP_020944374.1 - 3712 117
GenBank mRNA AF056489.1 - 6 - 7 567 117
AK344681.1 - 7 - 8 681 117

View Full Product Details