The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_214235.1 | NP_999400.1 | 3 - 4 | 276 | 68 |
XM_013977795.2 | XP_013833249.1 | - | 285 | 68 | |
XM_013977796.2 | XP_013833250.1 | - | 264 | 68 | |
GenBank mRNA | DQ355970.1 | - | 3 - 4 | 276 | 68 |
M31219.1 | - | 3 - 4 | 276 | 68 |