Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
MLN
Entrez Gene ID
397466
Gene Name
motilin
Gene Aliases
-
Chromosome Location
Chr.7: 30003501 - 30013064 on Build Sscrofa11.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_214235.1 NP_999400.1 4 - 5 393 49
XM_013977795.2 XP_013833249.1 - 402 49
XM_013977796.2 XP_013833250.1 - 381 49
GenBank mRNA DQ355970.1 - 4 - 5 393 49
M31219.1 - 4 - 5 393 49

View Full Product Details