Assay Design

The assay is designed to a sequence that does not exhibit typical exon-intron-exon gene structure, such as a fusion transcript, or an artificial, prokaryotic or mitochondrial sequence.

Gene Transcripts

Gene Symbol
EIF2AK2
Entrez Gene ID
397588
Gene Name
eukaryotic translation initiation factor 2 alpha kinase 2
Gene Aliases
PKR, PRKR
Chromosome Location
Chr.3: 103041029 - 103096741 on Build Sscrofa11.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_214319.1 NP_999484.1 15 - 16 1647 146
XM_021085862.1 XP_020941521.1 - 1780 146
GenBank mRNA AB104654.1 - 15 - 16 1647 146
AK236995.1 - 15 - 16 1652 146
AK393694.1 - 12 - 13 1230 146
AK396375.1 - 15 - 16 1763 146

View Full Product Details