Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
PPARG
Entrez Gene ID
397671
Gene Name
peroxisome proliferator activated receptor gamma
Gene Aliases
-
Chromosome Location
Chr.13: 68301566 - 68433951 on Build Sscrofa11.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_214379.1 NP_999544.1 5 - 6 829 141
XM_005669783.3 XP_005669840.1 - 925 141
XM_005669784.3 XP_005669841.1 - 986 141
XM_005669788.3 XP_005669845.3 - 842 141
XM_005669789.3 XP_005669846.1 - 913 141
XM_013981980.2 XP_013837434.1 - 759 141
XM_013981981.2 XP_013837435.1 - 1204 141
XM_013981982.2 XP_013837436.1 - 1102 141
GenBank mRNA AB097926.1 - 6 - 7 889 141
AB097927.1 - 5 - 6 815 141
AB097928.1 - 6 - 7 882 141
AB097929.1 - 5 - 6 808 141
AB097930.1 - 5 - 6 850 141
AF059245.1 - 5 - 6 829 141
AF103946.1 - 5 - 6 828 141
AJ006756.1 - 6 - 7 863 141
AJ006757.1 - 5 - 6 873 141
AK230961.1 - 5 - 6 141
AK389674.1 - 5 - 6 957 141
DQ437884.1 - 6 - 7 881 141
DQ437885.1 - 5 - 6 851 141
KM382175.1 - 4 - 5 726 141

View Full Product Details