Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
CYP2C33
Entrez Gene ID
403107
Gene Name
cytochrome P450 2C33
Gene Aliases
-
Chromosome Location
Chr.14: 111268751 - 111301748 on Build Sscrofa11.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_214414.1 NP_999579.1 2 - 3 380 121
GenBank mRNA AB052257.1 - 2 - 3 380 121
AK393722.1 - 2 - 3 394 121

Gene Symbol
LOC100739741
Entrez Gene ID
100739741
Gene Name
cytochrome P450 2C23-like
Gene Aliases
CYP2C33
Chromosome Location
Chr.14: 111234004 - 111262638 on Build Sscrofa11.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001315734.1 NP_001302663.1 2 - 3 702 121
GenBank mRNA AK233321.1 - 2 - 3 560 121
KP687253.1 - 2 - 3 710 121
KP687256.1 - 2 - 3 702 121

View Full Product Details