Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
TNNC1
Entrez Gene ID
100156435
Gene Name
troponin C1, slow skeletal and cardiac type
Gene Aliases
TNNC
Chromosome Location
Chr.13: 34591571 - 34594320 on Build Sscrofa11.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001130243.1 NP_001123715.1 3 - 4 211 71

View Full Product Details