Assay Design

The assay primers and probes lie within a single exon. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
CD14
Entrez Gene ID
100037938
Gene Name
CD14 molecule
Gene Aliases
-
Chromosome Location
Chr.2: 142348036 - 142349686 on Build Sscrofa11.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001097445.2 NP_001090914.2 2 - 2 1354 83
GenBank mRNA AB267810.1 - 2 - 2 1354 83
AK346107.1 - 1 - 1 1237 83
AK350223.1 - 2 - 2 1354 83
DQ357211.1 - 1 - 1 1017 83
EF051626.1 - 2 - 2 1242 83

View Full Product Details