Assay Design

The assay primers and probes lie within a single exon. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
IVL
Entrez Gene ID
407242
Gene Name
involucrin
Gene Aliases
-
Chromosome Location
Chr.4: 96656033 - 96658737 on Build Sscrofa11.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_214448.3 NP_999613.3 2 - 2 120 84
GenBank mRNA AK349671.1 - 2 - 2 118 84
AK349897.1 - 2 - 2 120 84

View Full Product Details