Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
CCNH
Entrez Gene ID
100310796
Gene Name
cyclin H
Gene Aliases
-
Chromosome Location
Chr.2: 94980938 - 95019053 on Build Sscrofa11.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001166313.1 NP_001159785.1 2 - 3 238 99
XM_005661543.3 XP_005661600.1 - 800 99
XM_005661544.3 XP_005661601.1 - 467 99
XM_013994968.2 XP_013850422.1 - 800 99
XM_013994969.2 XP_013850423.1 - 800 99
XM_013994970.2 XP_013850424.1 - 800 99
XM_013994971.2 XP_013850425.1 - 800 99
XM_013994972.2 XP_013850426.1 - 800 99
XM_021077281.1 XP_020932940.1 - 800 99
XM_021077311.1 XP_020932970.1 - 800 99
GenBank mRNA AB499892.1 - 2 - 3 238 99

View Full Product Details