Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
CDO1
Entrez Gene ID
100312964
Gene Name
cysteine dioxygenase type 1
Gene Aliases
-
Chromosome Location
Chr.2: 119927768 - 119940690 on Build Sscrofa11.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001167643.1 NP_001161115.1 1 - 2 173 100
GenBank mRNA AB529450.1 - 1 - 2 173 100
AK235587.1 - 1 - 2 437 100
AK395698.1 - 2 - 3 290 100
AY609406.1 - 1 - 2 403 100

View Full Product Details