Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
BMP6
Entrez Gene ID
100155536
Gene Name
bone morphogenetic protein 6
Gene Aliases
-
Chromosome Location
Chr.7: 5058817 - 5208868 on Build Sscrofa11.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001168001.1 NP_001161473.1 1 - 2 769 81
GenBank mRNA GU074521.1 - 1 - 2 769 81

View Full Product Details