Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
STK19
Entrez Gene ID
100144527
Gene Name
serine/threonine kinase 19
Gene Aliases
-
Chromosome Location
Chr.7: 24060226 - 24067398 on Build Sscrofa11.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001128450.1 NP_001121922.1 2 - 3 281 101
GenBank mRNA EU282371.1 - 3 - 4 954 101

View Full Product Details