Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
GBP2
Entrez Gene ID
100153137
Gene Name
guanylate binding protein 2, interferon-inducible
Gene Aliases
-
Chromosome Location
Chr.4: 127387222 - 127408729 on Build Sscrofa11.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001128474.1 NP_001121946.1 3 - 4 592 103
GenBank mRNA AK345662.1 - 2 - 3 379 103
AK393079.1 - 3 - 4 584 103
EU259888.1 - 3 - 4 592 103

Gene Symbol
LOC100523668
Entrez Gene ID
100523668
Gene Name
guanylate-binding protein 2-like
Gene Aliases
-
Chromosome Location
Chr.4: 127290026 - 127345677 on Build Sscrofa11.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_003125922.6 XP_003125970.6 - 794 103
XM_021090313.1 XP_020945972.1 - 801 103
XM_021090314.1 XP_020945973.1 - 787 103
XM_021090315.1 XP_020945974.1 - 465 103
XM_021090316.1 XP_020945975.1 - 779 103
XM_021090317.1 XP_020945976.1 - 606 103

View Full Product Details