Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
CYB5B
Entrez Gene ID
100286856
Gene Name
cytochrome b5 type B
Gene Aliases
-
Chromosome Location
Chr.6: 17522587 - 17558030 on Build Sscrofa11.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001159592.1 NP_001153064.1 3 - 4 373 60
GenBank mRNA AK352367.1 - 3 - 4 392 60
AY609739.1 - 3 - 4 373 60

View Full Product Details