Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
TRIM40
Entrez Gene ID
100294687
Gene Name
tripartite motif containing 40
Gene Aliases
-
Chromosome Location
Chr.7: 22702891 - 22717850 on Build Sscrofa11.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001160087.1 NP_001153559.1 4 - 5 1046 64
XM_021098154.1 XP_020953813.1 - 1176 64
XM_021098155.1 XP_020953814.1 - 1476 64

View Full Product Details