Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
LDHA
Entrez Gene ID
407245
Gene Name
lactate dehydrogenase A
Gene Aliases
LDH-A, LDH-M
Chromosome Location
Chr.2: 40814163 - 40827284 on Build Sscrofa11.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001172363.2 NP_001165834.1 3 - 4 350 69
XM_013994501.2 XP_013849955.2 - 1000 69
XM_021081651.1 XP_020937310.1 - 433 69
XM_021081655.1 XP_020937314.1 - 741 69
XM_021081659.1 XP_020937318.1 - 562 69
GenBank mRNA AK236433.1 - 3 - 4 388 69
AK343612.1 - 3 - 4 341 69
AK343811.1 - 3 - 4 339 69
AK391205.1 - 3 - 4 341 69
AK391208.1 - 3 - 4 338 69
AK391657.1 - 3 - 4 342 69
AK394718.1 - 1 - 2 107 69
AK395255.1 - 3 - 4 374 69
AK399432.1 - 3 - 4 342 69
AK399447.1 - 3 - 4 374 69
AK399466.1 - 3 - 4 341 69
AK399472.1 - 3 - 4 357 69
AK399544.1 - 3 - 4 341 69
AK399555.1 - 3 - 4 341 69
AK399560.1 - 5 - 6 550 69
FJ865398.1 - 3 - 4 351 69
U07178.1 - 3 - 4 337 69

View Full Product Details