Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
SPATA19
Entrez Gene ID
100415923
Gene Name
spermatogenesis associated 19
Gene Aliases
-
Chromosome Location
Chr.9: 60426620 - 60433458 on Build Sscrofa11.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001177920.1 NP_001171391.1 1 - 2 154 78
GenBank mRNA GU475012.1 - 1 - 2 154 78

View Full Product Details