Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
TAF11
Entrez Gene ID
100151814
Gene Name
TATA-box binding protein associated factor 11
Gene Aliases
-
Chromosome Location
Chr.7: 30799992 - 30811771 on Build Sscrofa11.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001184894.1 NP_001171823.1 3 - 4 505 92
GenBank mRNA AK236876.1 - 3 - 4 491 92
AK239635.1 - 3 - 4 470 92
AK398213.1 - 3 - 4 501 92
AY610074.1 - 3 - 4 472 92

View Full Product Details