Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
PMM1
Entrez Gene ID
100151790
Gene Name
phosphomannomutase 1
Gene Aliases
-
Chromosome Location
Chr.5: 6964999 - 6975904 on Build Sscrofa11.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001184895.1 NP_001171824.1 5 - 6 813 69
GenBank mRNA AK350347.1 - 5 - 6 514 69
AK392413.1 - 5 - 6 518 69
AK393586.1 - 5 - 6 520 69

View Full Product Details