Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
DNAJC14
Entrez Gene ID
100151876
Gene Name
DnaJ heat shock protein family (Hsp40) member C14
Gene Aliases
Jiv
Chromosome Location
Chr.5: 21264438 - 21273235 on Build Sscrofa11.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001185066.1 NP_001171995.1 4 - 5 1825 90
GenBank mRNA AK236843.1 - 2 - 3 157 90
AK350364.1 - 4 - 5 1790 90

View Full Product Details