Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
RANBP1
Entrez Gene ID
100151896
Gene Name
RAN binding protein 1
Gene Aliases
-
Chromosome Location
Chr.14: 51494258 - 51500420 on Build Sscrofa11.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001185070.1 NP_001171999.1 1 - 2 279 106
GenBank mRNA AK237655.1 - 1 - 2 279 106
AK239995.1 - 1 - 2 162 106

View Full Product Details