Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
RANBP1
Entrez Gene ID
100151896
Gene Name
RAN binding protein 1
Gene Aliases
-
Chromosome Location
Chr.14: 51494258 - 51500420 on Build Sscrofa11.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001185070.1 NP_001171999.1 2 - 3 416 65
XM_005670933.3 XP_005670990.1 - 570 65
GenBank mRNA AK237655.1 - 2 - 3 416 65
AK239995.1 - 2 - 3 299 65

View Full Product Details