Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
LIN52
Entrez Gene ID
100157325
Gene Name
lin-52 DREAM MuvB core complex component
Gene Aliases
-
Chromosome Location
Chr.7: 97382020 - 97491672 on Build Sscrofa11.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001185836.1 NP_001172765.1 5 - 6 302 85
GenBank mRNA AK231110.1 - 5 - 6 302 85

View Full Product Details