Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
TMEM38B
Entrez Gene ID
100153669
Gene Name
transmembrane protein 38B
Gene Aliases
-
Chromosome Location
Chr.1: 246973447 - 247024019 on Build Sscrofa11.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001190164.1 NP_001177093.1 5 - 6 713 107
GenBank mRNA AY609714.1 - 5 - 6 712 107

View Full Product Details