Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
MRO
Entrez Gene ID
100156956
Gene Name
maestro
Gene Aliases
-
Chromosome Location
Chr.1: 100342040 - 100368355 on Build Sscrofa11.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001190174.1 NP_001177103.1 1 - 2 95 71
XM_021100259.1 XP_020955918.1 - 494 71
XM_021100314.1 XP_020955973.1 - 282 71
XM_021100355.1 XP_020956014.1 - 768 71
GenBank mRNA AK234741.1 - 3 - 4 280 71
AK395077.1 - 3 - 4 283 71

View Full Product Details