Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
ECHS1
Entrez Gene ID
100156927
Gene Name
enoyl-CoA hydratase, short chain 1
Gene Aliases
-
Chromosome Location
Chr.14: 141340320 - 141348870 on Build Sscrofa11.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001190175.1 NP_001177104.1 1 - 2 179 114
GenBank mRNA AK232518.1 - 1 - 2 179 114
AK237861.1 - 1 - 2 127 114
AK346919.1 - 1 - 2 107 114
AK389542.1 - 1 - 2 333 114
AK392778.1 - 1 - 2 100 114

View Full Product Details