Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
KATNBL1
Entrez Gene ID
100154146
Gene Name
katanin regulatory subunit B1 like 1
Gene Aliases
C15orf29, C7H15orf29
Chromosome Location
Chr.7: 80287989 - 80383036 on Build Sscrofa11.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001190181.1 NP_001177110.1 9 - 10 1111 88
GenBank mRNA AK235777.1 - 9 - 10 944 88

View Full Product Details