Assay Design

The assay is designed to a sequence that does not exhibit typical exon-intron-exon gene structure, such as a fusion transcript, or an artificial, prokaryotic or mitochondrial sequence.

Gene Transcripts

Gene Symbol
CHMP4C
Entrez Gene ID
100154336
Gene Name
charged multivesicular body protein 4C
Gene Aliases
-
Chromosome Location
Chr.4: 54860481 - 54895229 on Build Sscrofa11.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001190190.1 NP_001177119.1 3 - 4 616 65
XM_021088560.1 XP_020944219.1 - 649 65
GenBank mRNA AK240538.1 - 3 - 4 616 65

View Full Product Details