Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
SNRPC
Entrez Gene ID
100154606
Gene Name
small nuclear ribonucleoprotein polypeptide C
Gene Aliases
-
Chromosome Location
Chr.7: 30708243 - 30723480 on Build Sscrofa11.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001190203.1 NP_001177132.1 5 - 6 400 85
GenBank mRNA AY610382.1 - 5 - 6 365 85
DY428718.1 - 3 - 4 301 85

View Full Product Details