Assay Design

The assay is designed to a sequence that does not exhibit typical exon-intron-exon gene structure, such as a fusion transcript, or an artificial, prokaryotic or mitochondrial sequence.

Gene Transcripts

Gene Symbol
GPD1
Entrez Gene ID
100153250
Gene Name
glycerol-3-phosphate dehydrogenase 1
Gene Aliases
cGPD
Chromosome Location
Chr.5: 16008179 - 16021676 on Build Sscrofa11.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001190240.1 NP_001177169.1 6 - 7 1097 94
XM_021090918.1 XP_020946577.1 - 1162 94
GenBank mRNA AK232843.1 - 6 - 7 1097 94

View Full Product Details