Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
MAP1LC3B
Entrez Gene ID
100462754
Gene Name
microtubule associated protein 1 light chain 3 beta
Gene Aliases
-
Chromosome Location
Chr.6: 1919879 - 1941069 on Build Sscrofa11.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001190290.1 NP_001177219.1 1 - 2 163 73
GenBank mRNA AK231642.1 - 1 - 2 140 73
AK389802.1 - 1 - 2 140 73
AK392018.1 - 1 - 2 141 73
HM046515.1 - 1 - 2 163 73

View Full Product Details