Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
STATH
Entrez Gene ID
100499199
Gene Name
statherin
Gene Aliases
-
Chromosome Location
Chr.8: 66944851 - 66953345 on Build Sscrofa11.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001193291.1 NP_001180220.1 5 - 6 271 71
GenBank mRNA GU332640.1 - 5 - 6 271 71

View Full Product Details