Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
C12H17orf75
Entrez Gene ID
100499565
Gene Name
chromosome 12 C17orf75 homolog
Gene Aliases
C17orf75
Chromosome Location
Chr.12: 42611934 - 42625300 on Build Sscrofa11.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001195120.1 NP_001182049.1 5 - 6 591 89
XM_021066070.1 XP_020921729.1 - 645 89
XM_021066071.1 XP_020921730.1 - 637 89
GenBank mRNA HM627269.1 - 5 - 6 591 89

View Full Product Details