Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
KAT7
Entrez Gene ID
100337667
Gene Name
lysine acetyltransferase 7
Gene Aliases
MYST2
Chromosome Location
Chr.12: 25970966 - 26005006 on Build Sscrofa11.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001195359.1 NP_001182288.1 13 - 14 1618 92
XM_005668865.3 XP_005668922.1 - 2102 92
XM_005668866.3 XP_005668923.1 - 2013 92
XM_005668867.3 XP_005668924.1 - 1538 92
GenBank mRNA GU373686.1 - 13 - 14 1618 92

View Full Product Details