Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
KAT7
Entrez Gene ID
100337667
Gene Name
lysine acetyltransferase 7
Gene Aliases
MYST2
Chromosome Location
Chr.12: 25970966 - 26005006 on Build Sscrofa11.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001195359.1 NP_001182288.1 10 - 11 1300 101
XM_005668865.3 XP_005668922.1 - 1784 101
XM_005668866.3 XP_005668923.1 - 1695 101
XM_005668867.3 XP_005668924.1 - 1220 101
GenBank mRNA GU373686.1 - 10 - 11 1300 101

View Full Product Details