Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
C4H1orf43
Entrez Gene ID
100518371
Gene Name
chromosome 4 C1orf43 homolog
Gene Aliases
C1orf43, NICE-3
Chromosome Location
Chr.4: 95520003 - 95530856 on Build Sscrofa11.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001204390.1 NP_001191319.1 1 - 2 323 90
GenBank mRNA AK231836.1 - 1 - 2 218 90
AK233670.1 - 1 - 2 291 90
AK234116.1 - 1 - 2 291 90
AK346595.1 - 1 - 2 291 90
AK351176.1 - 1 - 2 218 90
AK397935.1 - 1 - 2 366 90
AY609895.1 - 1 - 2 216 90
EU814887.1 - 1 - 2 292 90

View Full Product Details